Litiumaluminiumhydrid 1 M i THF, AcroSeal®
Leverantör: Thermo Scientific
Fara
Synonyms: Aluminium lithium hydride, Lithium tetrahydridoaluminate
199491000 199498000
199491000.EA 850 SEK
199491000. 199498000.
Litiumaluminiumhydrid 1 M i THF, AcroSeal®
Litiumaluminiumhydrid
Formel: H₄AlLi
MW: 37,95 g/mol
Flampunkt: –17 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011075
CAS nummer: 16853-85-3
EINECS: 240-877-9
UN: 1411
ADR: 4.3,I
Order Now
Specifikation testresultat
Appearance Turbid grey Solution
Titration Iodometric 3.9 to 4.5 % (as LiAlH4)