(±)-Methyl-2-chloropropionate, Sigma-Aldrich®
Leverantör: Merck
Varning
M-Clarity™ quality level = MQ200
8.20331.0250 8.20331.1000
8.20331.0250EA 729 SEK
8.20331.0250 8.20331.1000
(±)-Methyl-2-chloropropionate, Sigma-Aldrich®
(±)-Methyl-2-chloropropionate
Formel: C₄H₇ClO₂
Storage Temperature: Omgivande
MDL Number: MFCD00000863
CAS nummer: 17639-93-9
EINECS: 241-624-5
UN: 2933
ADR: 3,III
Order Now
Specifikation testresultat
Solubility (20 °C) practically insoluble
Molar Mass 122.55 g/mol
Boiling Point 127 - 129 °C
Vapor Pressure 30 hPa (20 °C)
Flash Point 36 °C DIN 51755 Part 1
Explosion Limit 2.8 - 13.9 %(V)
Density 1.13 g/cm3 (20 °C)
pH 6 - 7 (H2O, 20 °C)
Ignition Point 430 °C DIN 51794