(±)-2-Chloropropionyl chloride 96%
Leverantör: Thermo Fisher Scientific
Fara
B24503.14 B24503.22 B24503.36
B24503.14EA 775 SEK
B24503.14 B24503.22 B24503.36
(±)-2-Chloropropionyl chloride 96%
(±)-2-Chloropropionyl chloride
Purity: 96% CAS No.: 7623-09-8 Molecular Formula: CH3CHClCOCl Formula Weight: 126.97 Flash Point: 31°(88°F) Refractive Index: 1.4400 Boiling Point: 109-111° UN No.: 2920 MDL No.: MFCD00000862
Formel: C₃H₄Cl₂O
Kokpunkt: 109…111 °C
Densitet: 1,308
Flampunkt: 31 °C (88 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000862
CAS nummer: 7623-09-8
EINECS: 231-540-7
UN: 2920
ADR: 8,II