3,4-(Methylenedioxy)aniline 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 5-Amino-1,3-benzodioxole
171470100 171470500
ACRO171470100EA 610 SEK
ACRO171470100 ACRO171470500
3,4-(Methylenedioxy)aniline 97%
3,4-(Methylenedioxy)aniline
Formel: C₇H₇NO₂
Kokpunkt: 144 °C (16 mmHg)
Smältpunkt: 38…43 °C
Flampunkt: 142 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005832
CAS nummer: 14268-66-7
EINECS: 238-161-6
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Beige to brown to black-brown Crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 40 to 46 °C
GC ≥96.0 %
Titration with HClO4 96.0 to 104.0 %