(±)-Methyl-2-bromopropionate 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Methyl 2-bromopropanoate
173391000 173395000
ACRO173391000EA 875 SEK
ACRO173391000 ACRO173395000
(±)-Methyl-2-bromopropionate 99%
(±)-Methyl-2-bromopropionate
Formel: C₄H₇BrO₂
Kokpunkt: 143…145 °C (19 mmHg)
Densitet: 1,497 g/cm³
Flampunkt: 51 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000143
CAS nummer: 5445-17-0
EINECS: 226-642-3
UN: 2920
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4500 to 1.4530 (20°C, 589 nm)
GC ≥98.5 %