Triphenylphosphonium bromide ≥97%
Leverantör: Thermo Scientific
Fara
Synonyms: Triphenylphosphine hydrobromide
L14290.06 L14290.14
L14290.06EA 630 SEK
L14290.06 L14290.14
Triphenylphosphonium bromide ≥97%
Triphenylphosphonium bromide
Purity: 97% CAS No.: 6399-81-1 Molecular Formula: C18H15P.HBr Formula Weight: 343.21 Odour: Odourless Melting Point: ca 196° dec. Beilstein Reference: 3633387 UN No.: 3261 MDL No.: MFCD00035107
Formel: C₁₈H₁₆BrP
MW: 343,2 g/mol
Smältpunkt: ∼ 196 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00035107
CAS nummer: 6399-81-1
EINECS: 229-012-6
UN: 3261
ADR: 8,III