1-Amino-4-methylpiperazine 99%
Leverantör: Thermo Fisher Scientific
Varning
251390250 251391000
ACRO251390250EA 850 SEK
ACRO251390250 ACRO251391000
1-Amino-4-methylpiperazine 99%
1-Amino-4-methylpiperazine
Formel: C₅H₁₃N₃
Flampunkt: 62 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006154
CAS nummer: 6928-85-4
EINECS: 230-053-7
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4860 (20°C, 589 nm)
GC ≥98.5 %