Methyl propionylacetate 99+%
Leverantör: Thermo Fisher Scientific
Synonyms: Methyl 3-oxovalerate, Methyl 3-oxo-pentanoate
243160250 243161000 243165000
ACRO243160250EA 710 SEK
ACRO243160250 ACRO243161000 ACRO243165000
Methyl propionylacetate 99+%
Methyl propionylacetate
Formel: C₆H₁₀O₃
MW: 130.14 g/mol
Kokpunkt: 95…97 °C (48 mmHg)
Smältpunkt: –35 °C
Densitet: 1.03 g/cm³
Flampunkt: 71 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011705
CAS nummer: 30414-53-0
EINECS: 250-184-3
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4240 (20°C, 589 nm)
GC ≥99.0 %