Ethyl propionylacetate 98+%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Ethyl 3-oxopentanoate, Ethyl 3-oxovalerate
232910050 232910250
ACRO232910050EA 1100 SEK
ACRO232910050 ACRO232910250
Ethyl propionylacetate 98+%
Ethyl propionylacetate
Formel: C₇H₁₂O₃
Kokpunkt: 83…84 °C (12 mmHg)
Densitet: 1,01 g/cm³
Flampunkt: 77 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009317
CAS nummer: 4949-44-4
EINECS: 225-593-5
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4240 (20°C, 589 nm)
GC ≥98.0 %