Ammonium nickel(II) sulphate hexahydrate ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: Nickel ammonium sulphate hexahydrate
A18441.22 A18441.36
A18441.22EA 830 SEK
A18441.22 A18441.36
Ammonium nickel(II) sulphate hexahydrate ≥98%
Ammonium nickel(II) sulphate hexahydrate
Purity: 98% CAS No.: 7785-20-8 Molecular Formula: H8N2NiO8S2.6H2O Formula Weight: 394.99 (286.90anhy) Odour: Odourless Merck Reference: 14,6501 MDL No.: MFCD00150260
Formel: H₈N₂NiO₈S₂·6H₂O
MW: 394,99 g/mol
Densitet: 1,923
Storage Temperature: Omgivande
MDL Number: MFCD00150260
CAS nummer: 7785-20-8
EINECS: 239-793-5
ADR: 9,III
Merck Index: 14,6501