Ethyl palmitoleate
Leverantör: Cayman Chemical
Synonyms: Ethyl cis-9-Hexadecenoate, Palmitoleic acid ethyl ester, (Z)-Ethyl hexadec-9-enoate, cis-9-Hexadecenoic acid ethyl ester, Ethyl Palmitoleate
10008204-10 10008204-100 10008204-50 10008204-500
CAYM10008204-10EA 480 SEK
CAYM10008204-10 CAYM10008204-100 CAYM10008204-50 CAYM10008204-500
Ethyl palmitoleate
Ethyl palmitoleate
Formel: C₁₈H₃₄O₂
MW: 282.47 g/mol
MDL Number: MFCD00056314
CAS nummer: 56219-10-4
EINECS: 260-065-8