Adamantane ≥98%
Leverantör: Thermo Fisher Scientific
A14041.0B A14041.18 A14041.30
A14041.0BEA 10550 SEK
A14041.0B A14041.18 A14041.30
Adamantane ≥98%
Adamantane
Purity: 98% CAS No.: 281-23-2 Molecular Formula: C10H16 Formula Weight: 136.24 Melting Point: 266-268°(sealed tube) Beilstein Reference: 1901173 Merck Reference: 14,149 MDL No.: MFCD00074719
Formel: C₁₀H₁₆
Kokpunkt: 171 °C (1013 hPa)
Smältpunkt: 266…268 °C (sealed tube)
Densitet: 1,070
Storage Temperature: Omgivande
MDL Number: MFCD00074719
CAS nummer: 281-23-2
EINECS: 206-001-4
Merck Index: 14,149