2-Adamantanol ≥98%
Leverantör: Thermo Scientific
Synonyms: 2-Hydroxytricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decan-2-ol, 2-Tricyclo[3.3.1.1(3,7)]decanol
A10697.06 A10697.14
A10697.06EA 1025 SEK
A10697.06 A10697.14
2-Adamantanol ≥98%
2-Adamantanol
Purity: 98% CAS No.: 700-57-2 Molecular Formula: C10H16O Formula Weight: 152.24 Melting Point: 299-303°(sealed tube) Beilstein Reference: 1903957
Formel: C₁₀H₁₆O
MW: 152,24 g/mol
Smältpunkt: 299…303 °C (sealed tube)
Densitet: 1,07 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00074744
CAS nummer: 700-57-2
EINECS: 211-846-7