3-Hexylthiophene 98%

Leverantör: Thermo Scientific
Varning

367380050 367380250
ACRO367380050EA 1925 SEK
ACRO367380050 ACRO367380250
3-Hexylthiophene 98%
3-Hexylthiophene
Formel: C₁₀H₁₆S
MW: 168,3 g/mol
Kokpunkt: 65 °C (,4 mmHg)
Densitet: 0,936 g/cm³
Flampunkt: 37 °C
Storage Temperature: Omgivande
MDL Number: MFCD00143181
CAS nummer: 1693-86-3
UN: 1993
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4930 to 1.4990 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR