2-Octylthiophene ≥98%
Leverantör: Thermo Scientific
Synonyms:
2-n-Octylthiophene
A13672.06
A13672.14
A13672.18
A13672.06EA
1225
SEK
A13672.06
A13672.14
A13672.18
2-Octylthiophene ≥98%
2-Octylthiophene
Purity: 98%
CAS No.: 880-36-4
Molecular Formula: C12H20S
Formula Weight: 196.36
Flash Point:
Formel:
C₁₂H₂₀S MW: 196,36 g/mol Kokpunkt: 127…129 °C (12 mmHg) Densitet: 0,921 Flampunkt: >100 °C (212 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00041015 CAS nummer: 880-36-4 EINECS: 212-913-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...