2-Butylthiophene ≥98%
Leverantör: Thermo Scientific
Varning
A12740.06 A12740.14
A12740.06EA 1175 SEK
A12740.06 A12740.14
2-Butylthiophene ≥98%
2-Butylthiophene
Purity: 98+% CAS No.: 1455-20-5 Molecular Formula: C8H12S Formula Weight: 140.25 Flash Point: 56°(133°F) Refractive Index: 1.5020 Odour: Recognizable Boiling Point: 181-182° Beilstein Reference: 107046 UN No.: 1993 MDL No.: MFCD00022534
Formel: C₈H₁₂S
MW: 140,25 g/mol
Kokpunkt: 181…182 °C
Densitet: 0,953
Flampunkt: 56 °C (133 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00022534
CAS nummer: 1455-20-5
EINECS: 215-935-1
UN: 1993
ADR: 3,III