2-Nitrothiophene tech. 85%, Technical Grade
Leverantör: Thermo Fisher Scientific
Varning
A17464.06 A17464.14
A17464.06EA 885 SEK
A17464.06 A17464.14
2-Nitrothiophene tech. 85%, Technical Grade
2-Nitrothiophene
Purity: 85% CAS No.: 609-40-5 Molecular Formula: C4H3NO2S Formula Weight: 129.14 Flash Point: 94°(201°F) Melting Point: 43-45° Boiling Point: 224-225° MDL No.: MFCD00005425
Formel: C₄H₃NO₂S
Kokpunkt: 224…225 °C
Smältpunkt: 43…45 °C
Densitet: 1,3644
Flampunkt: 94 °C (201 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005425
CAS nummer: 609-40-5
EINECS: 210-190-9