2-Bromothiophene ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Thienyl bromide
A11959.0B A11959.18 A11959.30
A11959.0BEA 6850 SEK
A11959.0B A11959.18 A11959.30
2-Bromothiophene ≥98%
2-Bromothiophene
Purity: 98+% CAS No.: 1003-09-4 Molecular Formula: C4H3BrS Formula Weight: 163.04 Flash Point: 58°(136°F) Refractive Index: 1.5860 Melting Point: -10° Boiling Point: 149-152° Beilstein Reference: 104663 UN No.: 2929 MDL No.: MFCD00005417
Formel: C₄H₃BrS
MW: 163,04 g/mol
Kokpunkt: 149…152 °C
Smältpunkt: < –10 °C
Densitet: 1,701
Flampunkt: 58 °C (136 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005417
CAS nummer: 1003-09-4
EINECS: 213-699-4
UN: 2929
ADR: 6.1,II