2,4,6-Kollidin ≥99%
Leverantör: Thermo Scientific
Fara
A11058.AC A11058.AE A11058.AP
A11058.ACEA 550 SEK
A11058.AC A11058.AE A11058.AP
2,4,6-Kollidin ≥99%
2,4,6-Collidine
Purity: 99% CAS No.: 108-75-8 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 54°(129°F) Refractive Index: 1.4980 Odour: Unpleasant pH: 9-10 Melting Point: -43° Boiling Point: 171-172° Beilstein Reference: 107283 Merck Reference: 14,9718 UN No.: 1992 MDL No.: MFCD00006338
Formel: C₈H₁₁N
MW: 121,18 g/mol
Kokpunkt: 171…172 °C
Smältpunkt: –43 °C
Densitet: 0,916
Flampunkt: 54 °C (129 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006338
CAS nummer: 108-75-8
EINECS: 203-613-3
UN: 1992
ADR: 3,III
Merck Index: 14,9718