4-(3-Phenylpropyl)pyridine 98
Leverantör: Thermo Fisher Scientific
|
Varning
|
A13590.0B
A13590.18
A13590.30
A13590.0BEA
7275
SEK
A13590.0B
A13590.18
A13590.30
4-(3-Phenylpropyl)pyridine 98
4-(3-Phenylpropyl)pyridine
Purity: 98%
CAS No.: 2057-49-0
Molecular Formula: C14H15N
Formula Weight: 197.28
Flash Point: 140°(284°F)
Refractive Index: 1.5630
Boiling Point: 320-322°
Beilstein Reference: 142242
MDL No.: MFCD00047458
Formel:
C₁₄H₁₅N MW: 197.28 g/mol Kokpunkt: 320…322 °C Densitet: 0.853 Flampunkt: 140 °C (284 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00047458 CAS nummer: 2057-49-0 EINECS: 218-159-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...