3-(Trifluoromethyl)pyridine ≥97%
Leverantör: Thermo Fisher Scientific
Fara
L17280.03 L17280.06
L17280.03EA 2625 SEK
L17280.03 L17280.06
3-(Trifluoromethyl)pyridine ≥97%
3-(Trifluoromethyl)pyridine
Purity: 97% CAS No.: 3796-23-4 Molecular Formula: C6H4F3N Formula Weight: 147.10 Flash Point: 19°(66°F) Refractive Index: 1.4173 Boiling Point: 111-114° Beilstein Reference: 1563102 UN No.: 1993 MDL No.: MFCD00100032
Formel: C₆H₄F₃N
MW: 147.1 g/mol
Kokpunkt: 111…114 °C
Densitet: 1.28
Flampunkt: 19 °C (66 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00100032
CAS nummer: 3796-23-4
EINECS: 223-268-2
UN: 1993
ADR: 3 II