Pyridin, extra dry over Molecular Sieve 99.5%, AcroSeal®
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Azabenzene, Azine
364420010 364421000 364420025 364425000
364420010.EA 5000 SEK
364420010. 364421000. 364420025. 364425000.
Pyridin, extra dry over Molecular Sieve 99.5%, AcroSeal®
Pyridin
Formel: C₅H₅N
MW: 79.1 g/mol
Kokpunkt: 115…116 °C (760 mmHg)
Smältpunkt: –42 °C
Densitet: 0.978 g/cm³
Flampunkt: 17 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011732
CAS nummer: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3 II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.5 %