4-Aminopyridine 98%
Leverantör: Thermo Fisher Scientific
Fara
104570050 104570250 104571000 104570010
ACRO104570050EA 410 SEK
ACRO104570050 ACRO104570250 ACRO104571000 ACRO104570010
4-Aminopyridine 98%
4-Aminopyridine
Formel: C₅H₆N₂
Kokpunkt: 273 °C (760 mmHg)
Smältpunkt: 157…161 °C
Densitet: 1,25 g/cm³ (20 °C)
Flampunkt: 156 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006439
CAS nummer: 504-24-5
EINECS: 207-987-9
UN: 2671
ADR: 6.1,II
Merck Index: 13,03964
Order Now
Specifikation testresultat
Appearance White to light yellow to beige Crystalline powder or crystals or granules
Infrared spectrum Conforms
Melting point 157 to 161 °C
GC ≥97.5 %
Titration with HClO4 97.5 to 102.5 %