3-Fluoropyridine ≥98%
Leverantör: Thermo Fisher Scientific
Fara
A11693.06 A11693.14
A11693.06EA 1925 SEK
A11693.06 A11693.14
3-Fluoropyridine ≥98%
3-Fluoropyridine
Purity: 98% CAS No.: 372-47-4 Molecular Formula: C5H4FN Formula Weight: 97.09 Flash Point: 13°(55°F) Refractive Index: 1.4720 Boiling Point: 106-108° Beilstein Reference: 105702 UN No.: 1993 MDL No.: MFCD00006374
Formel: C₅H₄FN
MW: 97.09 g/mol
Kokpunkt: 106…108 °C
Densitet: 1.13
Flampunkt: 13 °C (55 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006374
CAS nummer: 372-47-4
EINECS: 206-755-4
UN: 1993
ADR: 3 II