2,3-Dichloropyridine ≥99%
Leverantör: Thermo Scientific
Fara
A12241.06 A12241.14 A12241.22
A12241.06EA 980 SEK
A12241.06 A12241.14 A12241.22
2,3-Dichloropyridine ≥99%
2,3-Dichloropyridine
Purity: 99% CAS No.: 2402-77-9 Molecular Formula: C5H3Cl2N Formula Weight: 147.99 Odour: Unpleasant Melting Point: 65-69° Beilstein Reference: 109811 UN No.: 2811 MDL No.: MFCD00006229
Formel: C₅H₃Cl₂N
MW: 147,99 g/mol
Kokpunkt: 202,5…203.5 °C (1013 hPa)
Smältpunkt: 65…69 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006229
CAS nummer: 2402-77-9
EINECS: 219-281-8
UN: 2811
ADR: 6.1,III