3-Bromopyridine
|
Varning
|
CC04110DA
MAYBCC04110DAEA
0
SEK
MAYBCC04110DA
3-Bromopyridine
3-Bromopyridine
Formel:
C₅H₄BrN MW: 158 g/mol Kokpunkt: 173 °C (1013 hPa) Smältpunkt: –27 °C Densitet: 1,62 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00006373 CAS nummer: 626-55-1 EINECS: 210-952-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...