3-Bromopyridine 99%
Leverantör: Thermo Scientific
Fara
107410250 107411000
ACRO107410250EA 1625 SEK
ACRO107410250 ACRO107411000
3-Bromopyridine 99%
3-Bromopyridine
Formel: C₅H₄BrN
MW: 158 g/mol
Kokpunkt: 173 °C (1013 hPa)
Smältpunkt: –27 °C
Densitet: 1,64 g/cm³
Flampunkt: 51 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006373
CAS nummer: 626-55-1
EINECS: 210-952-0
UN: 2929
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5680 to 1.5720 (20°C, 589 nm)
GC ≥98.5 %
UV at 280 nm A: ≤0.005
Color scale ≤10 APHA
Stabilizer 2 % Ethanol
Wavelength at 1 A.U. ≤205 nm
Residue after evaporation ≤2 ppm
Acidity/alkalinity ≤0.0002 meq/g