2-Bromopyridine ≥99%
Leverantör: Thermo Fisher Scientific
Fara
A13241.14 A13241.22 A13241.30 A13241.36
A13241.14EA 720 SEK
A13241.14 A13241.22 A13241.30 A13241.36
2-Bromopyridine ≥99%
2-Bromopyridine
Purity: 99% CAS No.: 109-04-6 Molecular Formula: C5H4BrN Formula Weight: 158.00 Flash Point: 97°(207°F) Refractive Index: 1.5720 Odour: Characteristic Boiling Point: 192-194° Beilstein Reference: 105789 UN No.: 2810 MDL No.: MFCD00006219
Formel: C₅H₄BrN
MW: 158 g/mol
Kokpunkt: 192…194 °C
Smältpunkt: –40.1 °C
Densitet: 1.625
Flampunkt: 97 °C (207 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006219
CAS nummer: 109-04-6
EINECS: 203-641-6
UN: 2810
ADR: 6.1 II