2,3-Dibromopyridine ≥97%
Leverantör: Thermo Fisher Scientific
Fara
L19527.14 L19527.03 L19527.06
L19527.14EA 10150 SEK
L19527.14 L19527.03 L19527.06
2,3-Dibromopyridine ≥97%
2,3-Dibromopyridine
Purity: 97% CAS No.: 13534-89-9 Molecular Formula: C5H3Br2N Formula Weight: 236.90 Melting Point: 58-60° Boiling Point: 249-250° MDL No.: MFCD00234014
Formel: C₅H₃Br₂N
MW: 236.89 g/mol
Kokpunkt: 249…250 °C
Smältpunkt: 58…60 °C
Storage Temperature: Omgivande
MDL Number: MFCD00234014
CAS nummer: 13534-89-9