Tetrahydrofuran, Supelco®
Leverantör: Merck
Fara
Synonyms: Butylene oxide, THF, Diethylene oxide
M-Clarity™ quality level = MQ100
87369-100ML 87369-500ML
87369-100MLEA 574 SEK
87369-100ML 87369-500ML
Tetrahydrofuran, Supelco®
Tetrahydrofuran
Formel: C₄H₈O
MW: 72.11 g/mol
Kokpunkt: 66
Smältpunkt: –108.5 °C
Densitet: 0.888 g/cm³ (20 °C)
Flampunkt: –21.5 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005356
CAS nummer: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=379da9efa44b1ccffbc0dd8673bc086bb5cd56de&vid=15b82ad4333ed2849c63ab34711d3472111e7b16&prid=web&clid=SAPDAM
Order Now
SPECIFIKATIONER
Environmentally Preferable