1-Bromo-2-methylnaphthalene tech. 90%, Technical Grade
Leverantör: Thermo Scientific
Varning
L05093.09 L05093.18
L05093.09EA 915 SEK
L05093.09 L05093.18
1-Bromo-2-methylnaphthalene tech. 90%, Technical Grade
1-Bromo-2-methylnaphthalene
Purity: 90% CAS No.: 2586-62-1 Molecular Formula: C11H9Br Formula Weight: 221.10 Flash Point: 110°(230°F) Refractive Index: 1.6490 Boiling Point: 295-296° Beilstein Reference: 2042531 MDL No.: MFCD00003871
Formel: C₁₁H₉Br
MW: 221,1 g/mol
Kokpunkt: 295…296 °C
Densitet: 1,428
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00003871
CAS nummer: 2586-62-1
EINECS: 219-966-1