1-Klornaftalen tech. 85% remainder 2-chloronaphthalene, Technical Grade
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Triisodecyl phosphite, α-Chloronaphthalene
A15029.0B A15029.18 A15029.30
A15029.0BEA 5950 SEK
A15029.0B A15029.18 A15029.30
1-Klornaftalen tech. 85% remainder 2-chloronaphthalene, Technical Grade
1-Klornaftalen
Purity: 85% CAS No.: 90-13-1 Molecular Formula: C10H7Cl Formula Weight: 162.62 Flash Point: 121°(249°F) Refractive Index: 1.6330 Melting Point: ca -8° Boiling Point: 259° Beilstein Reference: 970836 Merck Reference: 14,2149 MDL No.: MFCD00003874
Formel: C₁₀H₇Cl
MW: 162.62 g/mol
Kokpunkt: 259 °C
Smältpunkt: ∼–8 °C
Densitet: 1.194
Flampunkt: 121 °C (249 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00003874
CAS nummer: 90-13-1
EINECS: 201-967-3
UN: 2810
ADR: 6.1 III
Merck Index: 14,2149