Iron selenide ≥99.9%
Leverantör: Thermo Scientific
Fara
31112.18 31112.09
31112.18EA 20800 SEK
31112.18 31112.09
Iron selenide ≥99.9%
Iron selenide
Purity: 99.9% CAS No.: 1310-32-3 Molecular Formula: FeSe Formula Weight: 134.81 Physical Form: -40 Mesh Powder Sensitivity: Moisture Sensitive UN No.: 3283 MDL No.: MFCD00016085
Formel: FeSe
MW: 134,81 g/mol
Smältpunkt: 965 °C
Densitet: 4,72 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00016085
CAS nummer: 1310-32-3
EINECS: 215-177-1
UN: 3283
ADR: 6.1,III
Merck Index: 12,04103