Osmiumtetraoxid 99.8% (≥74.4% Os)
Leverantör: Thermo Scientific
Fara
Synonyms: Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid
12103.02 12103.03 12103.48
12103.02EA 3175 SEK
12103.02 12103.03 12103.48
Osmiumtetraoxid 99.8% (≥74.4% Os)
Osmiumtetraoxid
Purity: 99.8% CAS No.: 20816-12-0 Molecular Formula: OsO4 Formula Weight: 254.10 Physical Form: Crystalline Odour: Like chlorine Boiling Point: 130° Solubility: Soluble in chloroform, alcohol and ethers Merck Reference: 14,6893 UN No.: 2471 MDL No.: MFCD00011150
Oxidation, catalyst in Sharpless dihydroxylations
Formel: O₄Os
MW: 254,23 g/mol
Kokpunkt: 130 °C
Smältpunkt: 40,6 °C
Densitet: 1,04 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011150
CAS nummer: 20816-12-0
EINECS: 244-058-7
UN: 2471
ADR: 6.1,I
Merck Index: 14,6893