Krom(III)oxid, Patinal®
Leverantör: Merck
Synonyms:
Chromic oxide
1.02484.0100
1.02484.1000
1.02484.0100EA
0
SEK
1.02484.0100
1.02484.1000
Krom(III)oxid, Patinal®
Krom(III)oxid
Formel:
Cr₂O₃ MW: 151,99 g/mol Kokpunkt: 4000 °C (1013 hPa) Smältpunkt: 2435 °C Densitet: 5,22 g/cm³ (25 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00010949 CAS nummer: 1308-38-9 EINECS: 215-160-9 Merck Index: 13,02247 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...