Zirkonium(IV)oxid ≥99.7% (max. 75ppm Hf)
Leverantör: Thermo Fisher Scientific
Varning
41528.30 41528.18 41528.A1
41528.30EA 3425 SEK
41528.30 41528.18 41528.A1
Zirkonium(IV)oxid ≥99.7% (max. 75ppm Hf)
Zirconium(IV) oxide
Purity: 99.7% CAS No.: 1314-23-4 Molecular Formula: ZrO2 Formula Weight: 123.22 Physical Form: -325 Mesh Powder Density: 5.89 Melting Point: ca 2700° Boiling Point: ca 5000° Merck Reference: 14,10180 MDL No.: MFCD00011310
Formel: ZrO₂
Kokpunkt: ∼5000 °C
Smältpunkt: ∼2700 °C
Densitet: 5,89
Storage Temperature: Omgivande
MDL Number: MFCD00011310
CAS nummer: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180