Zirkonium(IV)oxid ≥99.978%, Puratronic®
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Zirconium dioxide, Zirconium Oxide
11141.14 11141.22 11141.36
11141.14EA 5775 SEK
11141.14 11141.22 11141.36
Zirkonium(IV)oxid ≥99.978%, Puratronic®
Zirconium(IV) oxide
Purity: 99.978% CAS No.: 1314-23-4 Molecular Formula: O2Zr Formula Weight: 123.22 Physical Form: Powder Density: 5.89 Appearance: White Odour: Odourless Melting Point: ca 2700° Boiling Point: ca 5000° Merck Reference: 14,10180 MDL No.: MFCD00011310
Formel: ZrO₂
MW: 123.22 g/mol
Kokpunkt: ∼5000 °C
Smältpunkt: ∼2700 °C
Densitet: 5.89
Storage Temperature: Omgivande
MDL Number: MFCD00011310
CAS nummer: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180