Zinkoxid, EMPROVE® EXPERT BP, Ph. Eur., JP, USP suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Varning
Synonyms: Zinci oxidum, Zinc (II) oxide
1.08846.9029 1.08846.1000
1.08846.9029EA 0 SEK
1.08846.9029 1.08846.1000
Zinkoxid, EMPROVE® EXPERT BP, Ph. Eur., JP, USP suitable for use as excipient, SAFC®
Zinkoxid
Formel: ZnO
Smältpunkt: 1975 °C
Densitet: 5,61 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011300
CAS nummer: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200