Zinkoxid ≥99%, nanopowder
Leverantör: Thermo Scientific
Varning
Synonyms: Zinci oxidum, Zinc (II) oxide
44533.09 44533.18
44533.09EA 2900 SEK
44533.09 44533.18
Zinkoxid ≥99%, nanopowder
Zinkoxid
Purity: 99% CAS No.: 1314-13-2 Molecular Formula: ZnO Formula Weight: 81.37 Physical Form: 20-30nm APS Powder Density: 5.606 Appearance: White Odour: Odourless Melting Point: 1975° Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300
Formel: ZnO
MW: 81,39 g/mol
Smältpunkt: 1975 °C
Densitet: 5,606
Storage Temperature: Omgivande
MDL Number: MFCD00011300
CAS nummer: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 14,10147