Zinkoxid 99.999%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Zinci oxidum, Zinc (II) oxide
315790250 315791000
ACRO315790250EA 2500 SEK
ACRO315790250 ACRO315791000
Zinkoxid 99.999%
Zinkoxid
Formel: ZnO
MW: 81.39 g/mol
Smältpunkt: 1975 °C
Densitet: 5.6 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00011300
CAS nummer: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9 III
REACH: 01-2119463881-32
Order Now
Specifikation testresultat
Appearance White to light yellow Powder and/or granules
Total trace metal impurities ≤0.001 %