Disilveroxid 99+%
Leverantör: Thermo Scientific
Fara
Synonyms: di-Silver oxide, Disilver oxide, Silver oxide
205800100 205800500 205802500
ACRO205800100EA 1825 SEK
ACRO205800100 ACRO205800500 ACRO205802500
Disilveroxid 99+%
Disilveroxid
Formel: Ag₂O
MW: 231,74 g/mol
Smältpunkt: 800…830 °C
Densitet: 7,14 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003404
CAS nummer: 20667-12-3
EINECS: 243-957-1
UN: 1479
ADR: 5.1,I
Merck Index: 12,08665
Order Now
Specifikation testresultat
Appearance Black Powder
Elemental Analysis ≥99.0 %
Nitrate (NO3) ≤50 ppm
Sulfate (SO4) ≤100 ppm
Aluminium (Al) ≤10 ppm
Iron (Fe) ≤30 ppm