Silvernitrat ≥99.995% Ag ≥ 63%, Premion®
Leverantör: Thermo Scientific
Fara
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
43087.22 43087.14 43087.06
43087.22EA 9400 SEK
43087.22 43087.14 43087.06
Silvernitrat ≥99.995% Ag ≥ 63%, Premion®
Silvernitrat
Purity: 99.995% CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Crystalline Density: 4.352 Appearance: White Odour: Odourless Melting Point: 212° Merck Reference: 14,8518 UN No.: 1479 MDL No.: MFCD00003414
Formel: AgNO₃
MW: 169,87 g/mol
Kokpunkt: 444 °C (1013 hPa)
Smältpunkt: 212 °C
Densitet: 4,352
Storage Temperature: Omgivande
MDL Number: MFCD00003414
CAS nummer: 7761-88-8
EINECS: 231-853-9
UN: 1479
ADR: 5.1,I
REACH: 01-2119513705-43
Merck Index: 14,8518
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=8eb5a021843d7bd34669386ef8c52ffd45ae3126&prid=web&clid=SAPDAM