Silverfluorid 99+%
Leverantör: Thermo Scientific
Fara
Synonyms: Silver monofluoride, Silver(I) fluoride, Argentous fluoride
210450050 210450250
ACRO210450050EA 2150 SEK
ACRO210450050 ACRO210450250
Silverfluorid 99+%
Silverfluorid
Formel: AgF
MW: 126,87 g/mol
Kokpunkt: 1150 °C (1013 hPa)
Smältpunkt: 435 °C
Densitet: 5,85 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00003410
CAS nummer: 7775-41-9
EINECS: 231-895-8
UN: 3260
ADR: 8,II
Merck Index: 12,08657
Order Now
Specifikation testresultat
Appearance Brown-yellow to rust brown Powder
Assay ≥99.0 %