Silverklorid 99+%, extra pure
Leverantör: Thermo Scientific
Varning
211270100 211270500 211272500
ACRO211270100EA 1525 SEK
ACRO211270100 ACRO211270500 ACRO211272500
Silverklorid 99+%, extra pure
Silver chloride
Formel: AgCl
MW: 143,32 g/mol
Kokpunkt: 1550 °C (760 mmHg)
Smältpunkt: 455 °C
Densitet: 5,56 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003399
CAS nummer: 7783-90-6
EINECS: 232-033-3
UN: 1759
ADR: 8,III
Merck Index: 13,08582
Order Now
Specifikation testresultat
Appearance White to pink-beige to grey Powder
Titration ≥99.0 %
Copper (Cu) ≤4 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤10 ppm