Silverbromid ≥99.998%, Premion®
Leverantör: Thermo Fisher Scientific
Synonyms: Argentous bromide
12110.06 12110.14 12110.22
12110.06EA 2700 SEK
12110.06 12110.14 12110.22
Silverbromid ≥99.998%, Premion®
Silverbromid
Purity: 99.998% CAS No.: 7785-23-1 Molecular Formula: AgBr Formula Weight: 187.78 Physical Form: Crystalline Density: 6.47 Appearance: Pale yellow Odour: Odourless Melting Point: 432° Merck Reference: 14,8506 UN No.: 3077 MDL No.: MFCD00003398
Formel: AgBr
Kokpunkt: 1300 °C (1013 hPa)
Smältpunkt: 432 °C
Densitet: 6,47
Storage Temperature: Omgivande
MDL Number: MFCD00003398
CAS nummer: 7785-23-1
EINECS: 232-076-8
UN: 3077
ADR: 9,III
Merck Index: 14,8506