Lantan(III)oxid ≥99.99% (REO, rare earth oxide basis), REacton®
Leverantör: Thermo Fisher Scientific
Varning
11264.30 11264.18 11264.A1
11264.30EA 4125 SEK
11264.30 11264.18 11264.A1
Lantan(III)oxid ≥99.99% (REO, rare earth oxide basis), REacton®
Lantan(III)oxid
Purity: 99.99% CAS No.: 1312-81-8 Molecular Formula: La2O3 Formula Weight: 325.82 Physical Form: Powder Density: 6.51 Melting Point: 2307° Boiling Point: 4200° Merck Reference: 14,5363 MDL No.: MFCD00011071
Formel: La₂O₃
MW: 325.81 g/mol
Kokpunkt: 4200 °C
Smältpunkt: 2307 °C
Densitet: 6.51
Storage Temperature: Omgivande
MDL Number: MFCD00011071
CAS nummer: 1312-81-8
EINECS: 215-200-5
Merck Index: 14,5363