Lantan(III)oxid 91%
Leverantör: Thermo Scientific
413021000 413022500
ACRO413021000EA 1375 SEK
ACRO413021000 ACRO413022500
Lantan(III)oxid 91%
Lantan(III)oxid
Formel: La₂O₃
MW: 325,81 g/mol
Kokpunkt: 4200 °C (1013 hPa)
Smältpunkt: 2315 °C
Densitet: 6,51 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011071
CAS nummer: 1312-81-8
EINECS: 215-200-5
Order Now
Specifikation testresultat
Appearance White to light grey Powder
Assay ≥89 %
Melting point 170 to 175 °C
GC ≥96.0 %
Titration with NaOH ≥96.0 %