Lanthanum(III) chloride, ultra dry ≥99.99%

Leverantör: Thermo Scientific
Fara

Synonyms: Lanthanum chloride, Lanthanum trichloride

Ampuled under argon

44322.06 44322.14
44322.06EA 1700 SEK
44322.06 44322.14
Lanthanum(III) chloride, ultra dry ≥99.99%
Lanthanum(III) chloride

Purity: 99.99% CAS No.: 10099-58-8 Molecular Formula: LaCl3 Formula Weight: 245.27 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.842 Appearance: White Odour: Odourless Melting Point: 860° Boiling Point: 1000° Merck Reference: 14,5363 UN No.: 3077 MDL No.: MFCD00011068

Formel: Cl₃La
MW: 245,26 g/mol
Kokpunkt: 1000 °C
Smältpunkt: 860 °C
Densitet: 3,842
Storage Temperature: Omgivande
MDL Number: MFCD00011068
CAS nummer: 10099-58-8
EINECS: 233-237-5
UN: 3077
ADR: 9,III
Merck Index: 14,5363

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR