Isokinolin 97%
Leverantör: Thermo Fisher Scientific
Fara
325031000 325035000
ACRO325031000EA 1250 SEK
ACRO325031000 ACRO325035000
Isokinolin 97%
Isokinolin
Formel: C₉H₇N
Kokpunkt: 242 °C (1013 hPa)
Smältpunkt: 24…28 °C
Densitet: 1,09 g/cm³
Flampunkt: 107 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00006898
CAS nummer: 119-65-3
EINECS: 204-341-8
UN: 2811
ADR: 6.1,II
Merck Index: 13,05242
Order Now
Specifikation testresultat
Appearance Colorless to white to yellow to light brown Fused solid or liquid
Infrared spectrum Conforms
Melting point 24 to 28 °C
GC ≥96.0 %