5-Aminoindan 97%

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: 5-Aminoindane

157350100 157350500
ACRO157350100EA 1375 SEK
ACRO157350100 ACRO157350500
5-Aminoindan 97%
5-Aminoindan
Formel: C₉H₁₁N
MW: 133.19 g/mol
Kokpunkt: 247…249 °C (745 mmHg)
Smältpunkt: 34…36 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003803
CAS nummer: 24425-40-9
EINECS: 246-241-7

Order Now

Specifikation testresultat

Appearance Brown Crystalline mass
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR